siRNA Details
Gene Symbol: NPTXR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014293.3 | NP_055108.2 | 5 | 2123 | ||
GenBank mRNA | AL161974.1 | - | 5 | 1436 | ||
AL162057.1 | - | 1 | 764 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
