siRNA Details
Gene Symbol: AIPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033054.2 | NP_001028226.1 | 5 | 1507 | ||
NM_001033055.2 | NP_001028227.1 | 5 | 1516 | |||
NM_001285399.2 | NP_001272328.1 | 6 | 1660 | |||
NM_001285400.2 | NP_001272329.1 | 6 | 1630 | |||
NM_001285401.2 | NP_001272330.1 | 6 | 1624 | |||
NM_001285402.1 | NP_001272331.1 | 6 | 1687 | |||
NM_014336.4 | NP_055151.3 | 6 | 1696 | |||
GenBank mRNA | AB593047.1 | - | 6 | 1674 | ||
AB593048.1 | - | 6 | 1685 | |||
AB593049.1 | - | 6 | 1639 | |||
AB593052.1 | - | 6 | 1617 | |||
AB593053.1 | - | 5 | 1473 | |||
AB593055.1 | - | 6 | 1661 | |||
AF148864.1 | - | 6 | 1784 | |||
AF525970.1 | - | 5 | 1507 | |||
BX537907.1 | - | 6 | 1590 | |||
BX647279.1 | - | 6 | 1661 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
