siRNA Details
Gene Symbol: IL17RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289905.1 | NP_001276834.1 | 5 | 606 | ||
NM_014339.6 | NP_055154.3 | 5 | 606 | |||
GenBank mRNA | BC011624.2 | - | 5 | 550 | 4053746 | |
BX334547.2 | - | 5 | 609 | |||
BX410574.2 | - | 5 | 522 | |||
CB044543.1 | - | 4 | 589 | |||
CT841520.1 | - | 5 | 529 | |||
JF305973.1 | - | 5 | 473 | |||
U58917.1 | - | 5 | 505 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
