siRNA Details
Gene Symbol: IL20RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278722.1 | NP_001265651.1 | 5, 6 | 854 | ||
NM_001278723.1 | NP_001265652.1 | 4, 5 | 994 | |||
NM_001278724.1 | NP_001265653.1 | 4, 5 | 928 | |||
NM_014432.3 | NP_055247.3 | 5, 6 | 1173 | |||
XM_006715506.2 | XP_006715569.1 | 4, 5 | 570 | |||
XM_011535904.2 | XP_011534206.1 | 4, 5 | 681 | |||
XM_017010954.1 | XP_016866443.1 | 5, 6 | 1362 | |||
XM_017010955.1 | XP_016866444.1 | 5, 6 | 822 | |||
GenBank mRNA | AF184971.1 | - | 5, 6 | 957 | ||
AJ420490.1 | - | 1, 2 | 148 | |||
AK098312.1 | - | 1, 2 | 763 | |||
AK297121.1 | - | 5, 6 | 816 | |||
AK315293.1 | - | 5, 6 | 961 | |||
AY358883.1 | - | 4, 5 | 508 | |||
BC113574.1 | - | 5, 6 | 1051 | 8322626 | ||
BC113602.1 | - | 5, 6 | 1051 | 8322654 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
