siRNA Details
Gene Symbol: SORCS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014978.2 | NP_055793.1 | 19 | 2796 | ||
XM_011539542.2 | XP_011537844.1 | 14 | 1547 | |||
GenBank mRNA | AB028982.1 | - | 19 | 2796 | ||
AK303009.1 | - | 6 | 860 | |||
BC117417.1 | - | 19 | 2612 | 40125968 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
