siRNA Details
Gene Symbol: JMJD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081461.1 | NP_001074930.1 | 2 | 607 | ||
NM_015167.2 | NP_055982.2 | 2 | 607 | |||
GenBank mRNA | AB011157.1 | - | 2 | 289 | ||
AB073711.1 | - | 2 | 283 | |||
AK021780.1 | - | 2 | 384 | |||
AK294816.1 | - | 2 | 487 | |||
AK314541.1 | - | 2 | 346 | |||
BC047003.1 | - | 2 | 398 | |||
BC066654.1 | - | 2 | 289 | 6064429 | ||
EF527406.1 | - | 2 | 592 | |||
EF527407.1 | - | 2 | 592 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
