siRNA Details
Gene Symbol: MOXD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015529.3 | NP_056344.2 | 8, 9 | 1422 | ||
XM_017010714.1 | XP_016866203.1 | 8, 9 | 1230 | |||
XM_017010715.1 | XP_016866204.1 | 8, 9 | 1068 | |||
GenBank mRNA | AK074879.1 | - | 8, 9 | 1137 | ||
AK226054.1 | - | 4, 5 | 519 | |||
AY007239.1 | - | 7, 8 | 1338 | |||
AY359094.1 | - | 8, 9 | 1346 | |||
BC018756.1 | - | 8, 9 | 1381 | 4841330 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
