siRNA Details
Gene Symbol: ADIPOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290553.1 | NP_001277482.1 | 4 | 493 | ||
NM_001290557.1 | NP_001277486.1 | 5 | 696 | |||
NM_001290629.1 | NP_001277558.1 | 5 | 582 | |||
NM_015999.5 | NP_057083.2 | 4 | 629 | |||
XM_017001407.1 | XP_016856896.1 | 5 | 580 | |||
GenBank mRNA | AF125179.1 | - | 4 | 497 | ||
AF125180.1 | - | 4 | 497 | |||
AF151803.1 | - | 4 | 505 | |||
AK001484.1 | - | 4 | 545 | |||
AK058114.1 | - | 4 | 545 | |||
AK222503.1 | - | 4 | 503 | |||
AY424279.1 | - | 4 | 465 | |||
BC001594.2 | - | 4 | 522 | 2988737 | ||
BC010743.1 | - | 4 | 516 | 3878067 | ||
BI597715.1 | - | 5 | 585 | |||
BX461217.2 | - | 5 | 582 | |||
DA523648.1 | - | 4 | 493 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
