siRNA Details
Gene Symbol: IL1RL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003856.3 | NP_003847.2 | 3 | 335 | ||
NM_016232.4 | NP_057316.3 | 3 | 369 | |||
NR_104167.1 | 3 | 335 | ||||
XM_006712839.3 | XP_006712902.1 | 3 | 335 | |||
XM_011512151.1 | XP_011510453.1 | 3 | 342 | |||
GenBank mRNA | AB012701.3 | - | 3 | 369 | ||
AB029084.1 | - | 3 | 297 | |||
AK291578.1 | - | 3 | 327 | |||
AL553735.3 | - | 3 | 321 | |||
BC030975.1 | - | 3 | 328 | 4770131 | ||
D12763.1 | - | 2 | 144 | |||
DA884586.1 | - | 3 | 336 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
