siRNA Details
Gene Symbol: GPR89A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097612.1 | NP_001091081.1 | 8, 9 | 900 | ||
NM_001097613.2 | NP_001091082.2 | 9, 10 | 946 | |||
NR_036541.1 | 8, 9 | 920 | ||||
XM_006711492.3 | XP_006711555.1 | 8, 9 | 870 | |||
XM_006711493.3 | XP_006711556.1 | 9, 10 | 878 | |||
XM_011509908.2 | XP_011508210.1 | 9, 10 | 916 | |||
XM_011509909.2 | XP_011508211.1 | 8, 9 | 1128 | |||
GenBank mRNA | AB097024.1 | - | 8, 9 | 841 | ||
AB097025.1 | - | 8, 9 | 841 | |||
AF132947.1 | - | 8, 9 | 825 | |||
AK021758.1 | - | 8, 9 | 890 | |||
AK301939.1 | - | 8, 9 | 747 | |||
AK316371.1 | - | 9, 10 | 858 | |||
AY358966.1 | - | 8, 9 | 875 | |||
BC003187.1 | - | 8, 9 | 803 | 3537949 | ||
BC067816.1 | - | 8, 9 | 859 | 5295014 | ||
BC146880.1 | - | 8, 9 | 821 | 9056808 | ||
BT006679.1 | - | 8, 9 | 726 | |||
KJ534847.1 | - | 8, 9 | 726 | |||
KJ535049.1 | - | 8, 9 | 746 |
Gene Symbol: GPR89B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016334.3 | NP_057418.1 | 8, 9 | 870 | ||
XM_005277401.3 | XP_005277458.1 | 8, 9 | 843 | |||
XM_005277402.4 | XP_005277459.1 | 8, 9 | 1007 | |||
XM_005277404.3 | XP_005277461.1 | 8, 9 | 863 | |||
XM_006711377.3 | XP_006711440.1 | 9, 10 | 906 | |||
XM_006711378.2 | XP_006711441.1 | 9, 10 | 909 | |||
XM_011509610.2 | XP_011507912.1 | 9, 10 | 903 | |||
XM_011509612.2 | XP_011507914.1 | 9, 10 | 864 | |||
XM_017001448.1 | XP_016856937.1 | 9, 10 | 1406 | |||
GenBank mRNA | AK299995.1 | - | 8, 9 | 841 | ||
AK302266.1 | - | 8, 9 | 862 | |||
CD102528.1 | - | 3, 4 | 235 | |||
KJ534850.1 | - | 8, 9 | 726 | |||
KJ534851.1 | - | 8, 9 | 657 | |||
KJ535070.1 | - | 9, 10 | 1054 | |||
U78723.1 | - | 8, 9 | 829 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
