siRNA Details
Gene Symbol: TPRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136053.2 | NP_001129525.1 | 7 | 918 | ||
NM_001142646.2 | NP_001136118.1 | 7 | 918 | |||
NR_073377.1 | 7 | 918 | ||||
XM_006713495.3 | XP_006713558.1 | 8 | 1099 | |||
XM_006713496.2 | XP_006713559.1 | 7 | 776 | |||
XM_006713497.3 | XP_006713560.1 | 8 | 1143 | |||
XM_011512422.1 | XP_011510724.1 | 7 | 692 | |||
XM_017005707.1 | XP_016861196.1 | 8 | 882 | |||
XM_017005708.1 | XP_016861197.1 | 8 | 912 | |||
XM_017005709.1 | XP_016861198.1 | 8 | 1013 | |||
GenBank mRNA | AB037108.1 | - | 6 | 842 | ||
AF289568.1 | - | 7 | 865 | |||
BC017058.1 | - | 7 | 687 | 3904719 | ||
BC050711.2 | - | 7 | 812 | 6168632 | ||
CD580484.2 | - | 7 | 653 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
