siRNA Details
Gene Symbol: TLR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016562.3 | NP_057646.1 | 3 | 1209 | ||
GenBank mRNA | AB445659.1 | - | 1 | 1070 | ||
AF240467.1 | - | 3 | 1205 | |||
AF245702.1 | - | 3 | 1203 | |||
AK313858.1 | - | 3 | 1220 | |||
AY358292.1 | - | 2 | 1154 | |||
BC033651.1 | - | 3 | 1220 | 5582912 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
