siRNA Details
Gene Symbol: CHST11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173982.1 | NP_001167453.1 | 2 | 619 | ||
NM_018413.5 | NP_060883.1 | 2 | 634 | |||
XM_017019369.1 | XP_016874858.1 | 3 | 141 | |||
GenBank mRNA | AB042326.1 | - | 2 | 522 | ||
AF239820.1 | - | 2 | 285 | |||
AJ269537.2 | - | 2 | 203 | |||
AJ289134.1 | - | 2 | 261 | |||
AK290923.1 | - | 2 | 575 | |||
AK313217.1 | - | 2 | 562 | |||
AY533204.1 | - | 154 | ||||
AY533205.1 | - | 152 | ||||
BC013315.1 | - | 3 | 431 | 4042813 |
