siRNA Details
Gene Symbol: TNFRSF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204458.1 | NP_001191387.1 | 4, 5 | 570 | ||
NM_001204459.1 | NP_001191388.1 | 2, 3 | 665 | |||
NM_018647.3 | NP_061117.2 | 4, 5 | 570 | |||
NM_148957.3 | NP_683760.1 | 4, 5 | 879 | |||
XM_005266445.2 | XP_005266502.1 | 4, 5 | 879 | |||
XM_005266446.1 | XP_005266503.1 | 4, 5 | 879 | |||
XM_011535146.1 | XP_011533448.1 | 5, 6 | 588 | |||
XM_017020651.1 | XP_016876140.1 | 5, 6 | 1091 | |||
XM_017020652.1 | XP_016876141.1 | 4, 5 | 570 | |||
GenBank mRNA | AB040434.1 | - | 3, 4 | 402 | ||
AF167555.1 | - | 4, 5 | 430 | |||
AF246998.1 | - | 3, 4 | 366 | |||
AF246999.1 | - | 3, 4 | 366 | |||
AK292874.1 | - | 4, 5 | 470 | |||
AK292891.1 | - | 3, 4 | 390 | |||
AK304290.1 | - | 2, 3 | 371 | |||
AY358888.1 | - | 4, 5 | 550 | |||
BC047321.1 | - | 4, 5 | 665 | 5248745 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
