siRNA Details
Gene Symbol: ACKR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020311.2 | NP_064707.1 | 2 | 1789 | ||
XM_005246097.2 | XP_005246154.1 | 2 | 1685 | |||
XM_005246098.3 | XP_005246155.1 | 3 | 1927 | |||
XM_017004516.1 | XP_016860005.1 | 2 | 1680 | |||
GenBank mRNA | AF030297.1 | - | 2 | 1637 | ||
BC008459.1 | - | 1 | 1186 | |||
BC036661.2 | - | 4 | 1986 | 5267661 | ||
U67784.1 | - | 1 | 1396 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
