siRNA Details
Gene Symbol: GPR107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136557.1 | NP_001130029.1 | 4, 5 | 622 | ||
NM_001136558.1 | NP_001130030.1 | 4, 5 | 622 | |||
NM_020960.4 | NP_066011.2 | 4, 5 | 622 | |||
XM_017014953.1 | XP_016870442.1 | 4, 5 | 583 | |||
XM_017014954.1 | XP_016870443.1 | 4, 5 | 583 | |||
GenBank mRNA | AB046844.1 | - | 4, 5 | 393 | ||
AF376725.1 | - | 4, 5 | 403 | |||
AK026244.1 | - | 4, 5 | 407 | |||
AK225456.1 | - | 4, 5 | 407 | |||
AK293092.1 | - | 4, 5 | 403 | |||
AL834359.1 | - | 3, 4 | 266 | |||
BC110518.2 | - | 4, 5 | 403 | 40033360 | ||
BC143655.1 | - | 4, 5 | 418 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
