siRNA Details
Gene Symbol: EDA2R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199687.2 | NP_001186616.1 | 2, 3 | 191 | ||
NM_001242310.1 | NP_001229239.1 | 1, 2 | 93 | |||
NM_001324201.1 | NP_001311130.1 | 2, 3 | 191 | |||
NM_001324206.1 | NP_001311135.1 | 2, 3 | 193 | |||
NM_021783.4 | NP_068555.1 | 2, 3 | 193 | |||
NR_136726.1 | 2, 3 | 191 | ||||
NR_136727.1 | 2, 3 | 193 | ||||
XM_011531000.1 | XP_011529302.1 | 2, 3 | 143 | |||
XM_011531001.1 | XP_011529303.1 | 2, 3 | 191 | |||
XM_011531002.1 | XP_011529304.1 | 2, 3 | 143 | |||
XM_011531003.2 | XP_011529305.1 | 2, 3 | 183 | |||
XM_011531004.1 | XP_011529306.1 | 2, 3 | 143 | |||
XM_011531006.2 | XP_011529308.1 | 2, 3 | 183 | |||
XM_011531007.2 | XP_011529309.1 | 2, 3 | 183 | |||
XM_017029703.1 | XP_016885192.1 | 2, 3 | 183 | |||
XM_017029704.1 | XP_016885193.1 | 2, 3 | 143 | |||
XM_017029706.1 | XP_016885195.1 | 2, 3 | 143 | |||
GenBank mRNA | AF298812.1 | - | 1, 2 | 83 | ||
AK296264.1 | - | 2, 3 | 143 | |||
AK314881.1 | - | 2, 3 | 159 | |||
AY152724.1 | - | 1, 2 | 83 | |||
AY358735.1 | - | 1, 2 | 86 | |||
AY358736.1 | - | 1, 2 | 86 | |||
BC034919.1 | - | 2, 3 | 140 | 5434545 | ||
BP249106.1 | - | 2, 3 | 159 | |||
DA215848.1 | - | 2, 3 | 192 | |||
DA574050.1 | - | 2, 3 | 143 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
