siRNA Details
Gene Symbol: IL21R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021798.3 | NP_068570.1 | 9 | 2953 | ||
NM_181078.2 | NP_851564.1 | 9 | 3306 | |||
NM_181079.4 | NP_851565.4 | 10 | 3447 | |||
XM_011545857.2 | XP_011544159.1 | 10 | 3483 | |||
XM_011545858.2 | XP_011544160.1 | 9 | 3292 | |||
XM_017023257.1 | XP_016878746.1 | 10 | 3436 | |||
GenBank mRNA | AA354979.1 | - | 1 | 104 | ||
BC004348.1 | - | 9 | 2918 | 3634520 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
