siRNA Details
Gene Symbol: FGFRL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004356.2 | NP_001004356.1 | 7 | 2694 | ||
NM_001004358.1 | NP_001004358.1 | 7 | 2577 | |||
NM_021923.3 | NP_068742.2 | 6 | 2567 | |||
XM_011513486.1 | XP_011511788.1 | 7 | 2698 | |||
GenBank mRNA | AF312678.1 | - | 2878 | |||
AJ277437.1 | - | 6 | 2567 | |||
AK000530.1 | - | 1 | 232 | |||
AK022597.1 | - | 1 | 1479 | |||
AY358303.1 | - | 2899 | ||||
BC015228.1 | - | 3 | 1978 | |||
BC036769.1 | - | 7 | 2706 | 5744684 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
