siRNA Details
Gene Symbol: MBOAT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146056.2 | NP_001139528.1 | 5 | 1214 | ||
NM_001146082.2 | NP_001139554.1 | 7 | 1420 | |||
NM_001146083.2 | NP_001139555.1 | 6 | 1293 | |||
NM_024298.4 | NP_077274.3 | 7 | 1420 | |||
XM_011527299.2 | XP_011525601.1 | 7 | 1624 | |||
XM_011527300.2 | XP_011525602.1 | 7 | 1015 | |||
XM_017027296.1 | XP_016882785.1 | 7 | 1588 | |||
GenBank mRNA | AK055908.1 | - | 7 | 1353 | ||
AK122789.1 | - | 8 | 1138 | |||
AK123290.1 | - | 1 | 1867 | |||
AK293199.1 | - | 5 | 824 | |||
AK298689.1 | - | 5 | 900 | |||
BC002512.2 | - | 6 | 870 | |||
BC003164.1 | - | 7 | 951 | 2958347 | ||
BC006309.2 | - | 7 | 1210 | 4110925 | ||
BC015857.1 | - | 6 | 857 | 4671596 | ||
EU016381.1 | - | 6 | 872 | |||
S82470.1 | - | 5 | 726 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
