siRNA Details
Gene Symbol: MBOAT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146056.2 | NP_001139528.1 | 6 | 2212 | ||
NM_001146083.2 | NP_001139555.1 | 7 | 2291 | |||
NM_024298.4 | NP_077274.3 | 8 | 2418 | |||
XM_011527299.2 | XP_011525601.1 | 8 | 2622 | |||
XM_011527300.2 | XP_011525602.1 | 8 | 2013 | |||
GenBank mRNA | AK055908.1 | - | 8 | 2351 | ||
AK122789.1 | - | 9 | 2132 | |||
AK123290.1 | - | 2 | 2865 | |||
AK298689.1 | - | 6 | 1898 | |||
BC002512.2 | - | 7 | 1868 | |||
BC003164.1 | - | 8 | 1949 | 2958347 | ||
BC015857.1 | - | 7 | 1855 | 4671596 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
