siRNA Details
Gene Symbol: TGS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024831.7 | NP_079107.6 | 1, 2 | 497 | ||
XM_006716485.2 | XP_006716548.1 | 1, 2 | 283 | |||
XM_017014004.1 | XP_016869493.1 | 1, 2 | 283 | |||
GenBank mRNA | AF286340.1 | - | 1, 2 | 587 | ||
AF432215.1 | - | 1, 2 | 245 | |||
AK314693.1 | - | 1, 2 | 271 | |||
AY028423.1 | - | 1, 2 | 281 | |||
BM853546.1 | - | 1, 2 | 497 | |||
DB082992.1 | - | 1, 2 | 285 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
