siRNA Details
Gene Symbol: TGS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317902.1 | NP_001304831.1 | 2 | 519 | ||
NM_024831.7 | NP_079107.6 | 3 | 584 | |||
XM_005251328.2 | XP_005251385.1 | 2 | 299 | |||
XM_006716485.2 | XP_006716548.1 | 3 | 370 | |||
XM_017014004.1 | XP_016869493.1 | 3 | 370 | |||
GenBank mRNA | AF286340.1 | - | 3 | 674 | ||
AF432215.1 | - | 3 | 332 | |||
AK314693.1 | - | 3 | 358 | |||
AY028423.1 | - | 3 | 368 | |||
BM853546.1 | - | 3 | 584 | |||
BX648548.1 | - | 2 | 282 | |||
DB082992.1 | - | 3 | 372 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
