siRNA Details
Gene Symbol: NECTIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030916.2 | NP_112178.2 | 9 | 2563 | ||
XM_005245508.3 | XP_005245565.1 | 8 | 2459 | |||
XM_011510021.2 | XP_011508323.1 | 9 | 2537 | |||
XM_011510022.2 | XP_011508324.1 | 8 | 2462 | |||
GenBank mRNA | AB105191.1 | - | 9 | 2520 | ||
AF160477.1 | - | 9 | 2399 | |||
AF218028.1 | - | 1 | 738 | |||
AK027753.1 | - | 9 | 2520 | |||
BC010423.1 | - | 9 | 2511 | 3610849 | ||
BX641083.1 | - | 1 | 1351 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
