siRNA Details
Gene Symbol: TLR10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017388.2 | NP_001017388.1 | 2 | 1763 | ||
NM_001195106.1 | NP_001182035.1 | 3 | 2080 | |||
NM_001195107.1 | NP_001182036.1 | 3 | 1952 | |||
NM_001195108.1 | NP_001182037.1 | 3 | 1983 | |||
NM_030956.3 | NP_112218.2 | 4 | 2269 | |||
XM_011513760.2 | XP_011512062.1 | 2 | 1657 | |||
XM_011513761.2 | XP_011512063.1 | 5 | 2350 | |||
XM_011513762.2 | XP_011512064.1 | 4 | 2030 | |||
GenBank mRNA | AB445680.1 | - | 1 | 1631 | ||
AF296673.1 | - | 3 | 2117 | |||
AK292026.1 | - | 2 | 1749 | |||
AK313468.1 | - | 2 | 1900 | |||
AY358300.1 | - | 2 | 1741 | |||
BC089406.1 | - | 2 | 2115 | 3066588 | ||
BC109111.2 | - | 1 | 2238 | 40035401 | ||
BC109112.2 | - | 2 | 1781 | 40035404 | ||
DQ011502.1 | - | 1 | 1631 | |||
DQ011503.1 | - | 1 | 1631 | |||
DQ011504.1 | - | 1 | 1631 | |||
DQ011505.1 | - | 1 | 1631 | |||
DQ011506.1 | - | 1 | 1631 | |||
DQ011507.1 | - | 1 | 1631 | |||
DQ011508.1 | - | 1 | 1631 | |||
DQ011509.1 | - | 1 | 1631 | |||
DQ011510.1 | - | 1 | 1631 | |||
DQ011511.1 | - | 1 | 1631 | |||
DQ011512.1 | - | 1 | 1631 | |||
DQ011513.1 | - | 1 | 1631 | |||
DQ011514.1 | - | 1 | 1631 | |||
DQ011515.1 | - | 1 | 1631 | |||
DQ011516.1 | - | 1 | 1631 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
