siRNA Details
Gene Symbol: HAVCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032782.4 | NP_116171.3 | 2 | 349 | ||
GenBank mRNA | AB924452.1 | - | 1 | 32 | ||
AF251707.1 | - | 2 | 221 | |||
AF450242.1 | - | 2 | 152 | |||
AF450243.1 | - | 2 | 152 | |||
AK027334.1 | - | 2 | 138 | |||
AK314406.1 | - | 2 | 156 | |||
AY069944.1 | - | 2 | 95 | |||
BC020843.1 | - | 2 | 204 | |||
BC063431.1 | - | 2 | 239 | 5229539 | ||
BP291914.1 | - | 2 | 349 | |||
JX049979.1 | - | 2 | 95 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
