siRNA Details
Gene Symbol: ITGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002211.3 | NP_002202.2 | 4 | 556 | ||
NM_033668.2 | NP_391988.1 | 3 | 335 | |||
NM_133376.2 | NP_596867.1 | 4 | 471 | |||
GenBank mRNA | AK291697.1 | - | 4 | 408 | ||
AL710802.1 | - | 3 | 347 | |||
BC020057.1 | - | 4 | 400 | 3924411 | ||
BX537407.1 | - | 4 | 410 | |||
DB292597.1 | - | 4 | 411 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
