siRNA Details
Gene Symbol: SORCS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013031.2 | NP_001013049.1 | 7 | 1280 | ||
NM_001206569.1 | NP_001193498.1 | 7 | 1280 | |||
NM_001206570.1 | NP_001193499.1 | 7 | 1280 | |||
NM_001206571.1 | NP_001193500.1 | 7 | 1280 | |||
NM_001206572.1 | NP_001193501.1 | 7 | 1280 | |||
NM_052918.4 | NP_443150.3 | 7 | 1280 | |||
XM_011539199.2 | XP_011537501.1 | 7 | 1137 | |||
XM_011539201.2 | XP_011537503.1 | 7 | 1137 | |||
XM_017015614.1 | XP_016871103.1 | 7 | 1137 | |||
XM_017015615.1 | XP_016871104.1 | 7 | 1137 | |||
XM_017015616.1 | XP_016871105.1 | 7 | 816 | |||
XM_017015617.1 | XP_016871106.1 | 7 | 615 | |||
GenBank mRNA | AB209142.1 | - | 7 | 1140 | ||
AF284756.1 | - | 7 | 1106 | |||
AK125464.1 | - | 7 | 1280 | |||
AK126905.1 | - | 7 | 644 | |||
AK289797.1 | - | 7 | 1281 | |||
AY099452.1 | - | 7 | 1106 | |||
AY099453.1 | - | 7 | 1106 | |||
BC131597.1 | - | 7 | 1106 | 40114197 | ||
EU281343.1 | - | 7 | 1106 | |||
EU281344.1 | - | 7 | 1106 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
