siRNA Details
Gene Symbol: IL22RA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052962.2 | NP_443194.1 | 3 | 407 | ||
NM_181309.1 | NP_851826.1 | 3 | 407 | |||
NM_181310.1 | NP_851827.1 | 3 | 407 | |||
GenBank mRNA | AJ297262.1 | - | 2 | 127 | ||
AJ313161.1 | - | 3 | 409 | |||
AJ313162.1 | - | 3 | 409 | |||
AY040566.1 | - | 3 | 380 | |||
AY040567.1 | - | 3 | 380 | |||
AY040568.1 | - | 3 | 380 | |||
AY044429.1 | - | 3 | 342 | |||
AY358111.1 | - | 3 | 405 | |||
AY358737.1 | - | 3 | 343 | |||
BC125167.1 | - | 2 | 169 | 40121609 | ||
BC125168.1 | - | 2 | 169 | 40121612 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
