siRNA Details
Gene Symbol: PSAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021154.4 | NP_066977.1 | 8 | 1872 | ||
NM_058179.3 | NP_478059.1 | 9 | 2010 | |||
GenBank mRNA | AK025460.1 | - | 9 | 1944 | ||
BC000971.1 | - | 9 | 1932 | 3447262 | ||
BC004863.2 | - | 9 | 1941 | 3938160 | ||
BC016645.2 | - | 9 | 1920 | 4342332 | ||
BC018129.1 | - | 9 | 1941 | 3883843 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Matching Taqman Gene Expression Assay:
Gene Ontology Categories:
