siRNA Details
Gene Symbol: HLA-DQA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002122.3 | NP_002113.2 | 5 | 867 | ||
GenBank mRNA | AB209628.1 | - | 3 | 1365 | ||
AF533896.1 | - | 1 | 258 | |||
AF533907.1 | - | 2 | 91 | |||
AF533913.1 | - | 2 | 91 | |||
AF533939.1 | - | 1 | 258 | |||
AK130811.1 | - | 5 | 867 | |||
M17846.1 | - | 3 | 462 | |||
M20431.1 | - | 5 | 748 | |||
X00452.1 | - | 5 | 836 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
