siRNA Details
Gene Symbol: CSF3R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000760.3 | NP_000751.1 | 17 | 3273 | ||
NM_156039.3 | NP_724781.1 | 17 | 3354 | |||
NM_172313.2 | NP_758519.1 | 18 | 2853 | |||
XM_005270493.1 | XP_005270550.1 | 17 | 3270 | |||
XM_011540748.2 | XP_011539050.1 | 16 | 2879 | |||
XM_011540749.1 | XP_011539051.1 | 17 | 3351 | |||
XM_011540750.1 | XP_011539052.1 | 12 | 2184 | |||
XM_017000370.1 | XP_016855859.1 | 16 | 2916 | |||
GenBank mRNA | AB209656.1 | - | 9 | 5939 | ||
BC053585.1 | - | 17 | 2866 | 5217305 | ||
M59818.1 | - | 17 | 2858 | |||
M59819.1 | - | 17 | 2770 | |||
M59820.1 | - | 17 | 2939 | |||
X55720.1 | - | 18 | 2433 | |||
X55721.1 | - | 17 | 2854 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
