siRNA Details
Gene Symbol: IFNLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170743.3 | NP_734464.1 | 2 | 188 | ||
NM_173064.2 | NP_775087.1 | 2 | 188 | |||
NM_173065.2 | NP_775088.1 | 2 | 188 | |||
GenBank mRNA | AF439325.1 | - | 2 | 161 | ||
AJ534330.1 | - | 2 | 174 | |||
AJ534331.1 | - | 2 | 174 | |||
AK160364.1 | - | 2 | 164 | |||
AY129151.1 | - | 2 | 161 | |||
AY129152.1 | - | 2 | 161 | |||
AY129153.1 | - | 2 | 161 | |||
BC101975.1 | - | 2 | 174 | |||
BC139722.1 | - | 2 | 174 | |||
BC140872.1 | - | 2 | 237 | 9021740 | ||
CN289861.1 | - | 2 | 188 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
