siRNA Details
Gene Symbol: GPR119 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178471.2 | NP_848566.1 | 1 | 854 | ||
GenBank mRNA | AY288416.1 | - | 1 | 764 | ||
BC095502.1 | - | 1 | 788 | 30706606 | ||
BC101166.1 | - | 1 | 764 | 40016011 | ||
BC101167.1 | - | 1 | 764 | 40016012 | ||
BC101168.2 | - | 1 | 764 | 40016013 | ||
BC126179.1 | - | 1 | 854 | 8991895 | ||
BC126181.1 | - | 1 | 854 | 8991897 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
