siRNA Details
Gene Symbol: CD300E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181449.2 | NP_852114.2 | 4 | 818 | ||
GenBank mRNA | AF395839.1 | - | 4 | 722 | ||
AK298031.1 | - | 4 | 717 | |||
AK303545.1 | - | 4 | 818 | |||
BX648376.1 | - | 4 | 800 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
