siRNA Details
Gene Symbol: QRFPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198179.2 | NP_937822.2 | 6 | 1483 | ||
GenBank mRNA | AB109629.1 | - | 6 | 1104 | ||
BC128132.1 | - | 7 | 1235 | |||
BC128133.1 | - | 6 | 1104 | 40112420 | ||
JF810892.1 | - | 6 | 1104 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
