siRNA Details
Gene Symbol: RARRES1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002888.3 | NP_002879.2 | 2 | 370 | ||
NM_206963.1 | NP_996846.1 | 2 | 370 | |||
XM_005247686.4 | XP_005247743.3 | 2 | 798 | |||
GenBank mRNA | AK130079.1 | - | 2 | 341 | ||
BC029640.1 | - | 2 | 306 | 4776506 | ||
BM919188.1 | - | 2 | 496 | |||
U27185.1 | - | 2 | 335 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
