siRNA Details
Gene Symbol: OR10A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207186.2 | NP_997069.2 | 1 | 285 | ||
GenBank mRNA | AF209506.1 | - | 1 | 262 | ||
BC137024.1 | - | 1 | 285 | 9021019 | ||
BC137027.1 | - | 1 | 285 | 9021022 |
