siRNA Details
Gene Symbol: IL12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002187.2 | NP_002178.2 | 8 | 2324 | ||
GenBank mRNA | M65290.1 | - | 7 | 2295 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
