siRNA Details
Gene Symbol: ADAM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK301147.1 | - | 20 | 2392 | ||
BC064500.1 | - | 22 | 2480 | |||
CR997306.1 | - | 4 | 559 | |||
D26579.1 | - | 23 | 2623 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
