siRNA Details
Gene Symbol: ALPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052947.3 | NP_443179.3 | 8, 9 | 5877 | ||
XM_011525801.2 | XP_011524103.1 | 8, 9 | 6110 | |||
XM_011525802.2 | XP_011524104.1 | 6, 7 | 6497 | |||
XM_017025535.1 | XP_016881024.1 | 8, 9 | 6111 | |||
XM_017025536.1 | XP_016881025.1 | 8, 9 | 6111 | |||
XM_017025537.1 | XP_016881026.1 | 8, 9 | 6112 | |||
XM_017025538.1 | XP_016881027.1 | 5, 6 | 4995 | |||
XM_017025539.1 | XP_016881028.1 | 5, 6 | 3743 | |||
GenBank mRNA | AL832018.2 | - | 5, 6 | 4209 | ||
AY044450.1 | - | 5, 6 | 3944 | |||
BX647446.1 | - | 4, 5 | 3355 | |||
BX647639.1 | - | 5, 6 | 4743 | |||
BX647796.1 | - | 8, 9 | 5864 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
