siRNA Details
Gene Symbol: PLAU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | A18397.1 | - | 12 | 1880 | ||
A35395.1 | - | 11 | 2213 | |||
BC013575.2 | - | 11 | 2232 | 3890980 | ||
M15476.1 | - | 11 | 2220 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
