siRNA Details
Gene Symbol: UEVLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040697.2 | NP_001035787.1 | 12 | 1431 | ||
NM_001261382.1 | NP_001248311.1 | 11 | 1365 | |||
NM_001261383.1 | NP_001248312.1 | 10 | 1241 | |||
NM_001261384.1 | NP_001248313.1 | 10 | 1182 | |||
NM_001261385.1 | NP_001248314.1 | 11 | 1264 | |||
NM_001261386.1 | NP_001248315.1 | 7 | 772 | |||
NM_018314.4 | NP_060784.3 | 11 | 1307 | |||
XM_006718255.2 | XP_006718318.1 | 11 | 1260 | |||
XM_011520201.2 | XP_011518503.1 | 8 | 3262 | |||
XM_017017985.1 | XP_016873474.1 | 11 | 1358 | |||
XM_017017986.1 | XP_016873475.1 | 10 | 1114 | |||
GenBank mRNA | AA160640.1 | - | 3 | 247 | ||
AF503350.1 | - | 11 | 1267 | |||
AK001930.1 | - | 11 | 1264 | |||
AK295501.1 | - | 10 | 1110 | |||
AK296835.1 | - | 11 | 1325 | |||
AK301537.1 | - | 11 | 1307 | |||
AK316191.1 | - | 11 | 1238 | |||
AL134932.1 | - | 1 | 72 | |||
BC064566.1 | - | 10 | 1181 | 5576632 |
