siRNA Details
Gene Symbol: STK35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080836.3 | NP_543026.2 | 2, 3 | 900 | ||
GenBank mRNA | AF195026.1 | - | 1, 2 | 501 | ||
AK314728.1 | - | 2, 3 | 752 | |||
BC017340.2 | - | 1, 2 | 483 | |||
BC140883.1 | - | 1, 2 | 601 | 9021751 | ||
BC140884.1 | - | 1, 2 | 601 | 9021752 | ||
GQ281297.1 | - | 2, 3 | 900 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
