siRNA Details
Gene Symbol: CACNA1I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021096.3 | NP_066919.2 | 9 | 1516 | ||
GenBank mRNA | AF129133.1 | - | 9 | 1516 | ||
AF142567.1 | - | 9 | 1516 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
