siRNA Details
Gene Symbol: USP48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032236.5 | NP_115612.4 | 12 | 1762 | ||
XM_005246009.2 | XP_005246066.1 | 12 | 1766 | |||
XM_006710951.2 | XP_006711014.2 | 12 | 1763 | |||
XM_011542262.2 | XP_011540564.1 | 12 | 1656 | |||
XM_011542263.2 | XP_011540565.1 | 12 | 1659 | |||
XM_011542264.1 | XP_011540566.1 | 11 | 1356 | |||
XM_011542265.2 | XP_011540567.1 | 13 | 2322 | |||
XM_011542267.2 | XP_011540569.1 | 12 | 1675 | |||
XM_011542268.2 | XP_011540570.1 | 4 | 375 | |||
XM_017002496.1 | XP_016857985.1 | 12 | 1766 | |||
XM_017002497.1 | XP_016857986.1 | 4 | 420 | |||
GenBank mRNA | AF502942.1 | - | 12 | 1664 | ||
AK021830.1 | - | 1 | 1098 | |||
AK026707.1 | - | 1 | 428 | |||
AK074247.1 | - | 12 | 1656 | |||
AK315005.1 | - | 12 | 1542 | |||
BC007326.2 | - | 3 | 330 | |||
BC067261.1 | - | 12 | 1730 | |||
BC103724.1 | - | 12 | 1704 | |||
BC104896.1 | - | 12 | 1558 | 8143899 | ||
BC143343.1 | - | 12 | 1558 | 9051848 | ||
BC143349.1 | - | 12 | 1558 | 9051854 | ||
BC143354.1 | - | 12 | 1555 | 9051859 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
