siRNA Details
Gene Symbol: HSD11B1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267868.1 | NP_001254797.1 | 9 | 1631 | ||
NM_001267869.1 | NP_001254798.1 | 6 | 1318 | |||
NM_001267870.1 | NP_001254799.1 | 5 | 1158 | |||
NM_001267871.1 | NP_001254800.1 | 6 | 1087 | |||
NM_198533.2 | NP_940935.1 | 8 | 1330 | |||
NM_198704.2 | NP_941993.1 | 6 | 1155 | |||
NM_198705.2 | NP_941994.1 | 6 | 1250 | |||
NM_198706.2 | NP_941995.1 | 8 | 1493 | |||
NM_198707.2 | NP_941996.1 | 7 | 1430 | |||
NM_198708.2 | NP_941997.1 | 7 | 1381 | |||
GenBank mRNA | AK308206.1 | - | 9 | 1372 | ||
AY268351.1 | - | 6 | 991 | |||
AY268352.1 | - | 8 | 1194 | |||
AY268353.1 | - | 7 | 1154 | |||
AY268354.1 | - | 7 | 1082 | |||
AY313894.1 | - | 8 | 1059 | |||
AY313895.1 | - | 6 | 896 | |||
AY313896.1 | - | 6 | 828 | |||
BC053546.1 | - | 7 | 1011 | |||
BC072434.1 | - | 6 | 1067 | |||
BC094798.1 | - | 6 | 1074 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
