siRNA Details
Gene Symbol: NRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244972.1 | NP_001231901.1 | 17 | 3534 | ||
NM_001244973.1 | NP_001231902.1 | 17 | 3531 | |||
NM_003873.5 | NP_003864.4 | 17 | 3552 | |||
NR_045259.1 | 16 | 3349 | ||||
XM_006717521.1 | XP_006717584.1 | 17 | 3329 | |||
XM_006717522.1 | XP_006717585.1 | 16 | 3278 | |||
XM_006717523.2 | XP_006717586.1 | 16 | 3275 | |||
XM_006717524.1 | XP_006717587.1 | 16 | 3224 | |||
XM_006717525.1 | XP_006717588.1 | 16 | 3221 | |||
XM_017016865.1 | XP_016872354.1 | 16 | 3254 | |||
XM_017016866.1 | XP_016872355.1 | 16 | 3146 | |||
GenBank mRNA | AB209641.1 | - | 17 | 3180 | ||
AK292822.1 | - | 17 | 3200 | |||
AL833608.1 | - | 16 | 3080 | |||
BX510902.1 | - | 17 | 3216 | |||
BX648250.1 | - | 17 | 3500 | |||
CR749333.1 | - | 17 | 3553 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
