siRNA Details
Gene Symbol: SREBF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004176.4 | NP_004167.3 | 1, 2 | 282 | ||
XM_005256772.4 | XP_005256829.1 | 1, 2 | 288 | |||
GenBank mRNA | AB373958.1 | - | 1, 2 | 88 | ||
BC057388.1 | - | 1, 2 | 273 | 5786483 | ||
BI906407.1 | - | 280 | ||||
U00968.1 | - | 1, 2 | 254 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
