siRNA Details
Gene Symbol: FPGS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110170.1 | 4, 5 | 449 | |||
XM_011518439.2 | XP_011516741.1 | 1, 2 | 14 | |||
GenBank mRNA | AK315523.1 | - | 4, 5 | 427 | ||
BT006639.1 | - | 4, 5 | 382 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
